Learn More
Sino Biological Anti-IL-20 Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000751004
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-20 (rh IL-20; Catalog#13060-HNAE; Q9NYY1; Leu25-Glu176). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-51004 Supplier Sino Biological Supplier No. 13060MM03T100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-20 (rh IL-20; Catalog#13060-HNAE; Q9NYY1; Leu25-Glu176). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.