Learn More
Sino Biological Anti-IL-21 Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000786592
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL-21 (rR IL-21; Catalog#80200-R08B; NP_001102413.1; Met1-Ser146). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-86592 Supplier Sino Biological Supplier No. 80200MM01100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL-21 (rR IL-21; Catalog#80200-R08B; NP_001102413.1; Met1-Ser146). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.