Learn More
Sino Biological Anti-IL1R1 Antibody (Biotin), Mouse Monoclonal 20 µg
Catalog No. 5000785488
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL-1R1 / CD121a (rR IL-1R1 / CD121a; Catalog#80028-R08H; NP_037255.3; Met 1-Lys 352). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-007-85488 Supplier Sino Biological Supplier No. 80028MM08B20
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL-1R1 / CD121a (rR IL-1R1 / CD121a; Catalog#80028-R08H; NP_037255.3; Met 1-Lys 352). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.