Learn More
CPC Scientific H-Lys-Ser-Arg-Ile-Val-Pro-Ala-Ile-Pro-Val-Ser-Leu-Leu-NH2 1MG
SDP
Supplier:  CPC Scientific ATIF001A
Encompass
The anti-infective peptide IDR-1 can selectively act on the host to stimulate the innate immune response. IDR-1 is believed to enhancing monocyte chemokines and reducing pro-inflammatory cytokines (decreased TNF and IL-6 and increased IL-10).SEQUENCE: H-Lys-Ser-Arg-Ile-Val-Pro-Ala-Ile-Pro-Val-Ser-Leu-Leu-NH2ONE-LETTER SEQUENCE: KSRIVPAIPVSLL-NH2MOLECULAR FORMULA: C65H118N18O15MOLECULAR WEIGHT: 1391.76STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [940291-10-1]SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:M.G.Scott et al., Nat. Biotechnol., 25, 465 (2007)
SKU(s): ATIF-001A, ATIF-001B
Catalog No. 50-210-8801