Learn More
Apexbio Technology LLC Anti-Inflammatory Peptide 1 118850-71-8 5mg
SDP
Supplier:  Apexbio Technology LLC A10085
Encompass_Preferred
Anti-Inflammatory Peptide 1 (CAS 118850-71-8) is a synthetic nonapeptide (H-Met-Gln-Met-Lys-Lys-Val-Leu-Asp-Ser-OH C45H82N12O14S2) structurally related to regions of uteroglobin and lipocortin I This peptide demonstrates significant inhibitory activity against phospholipase A2 (PLA2) an enzyme whose elevated expression and activity are associated with inflammatory responses By suppressing PLA2 Anti-Inflammatory Peptide 1 effectively reduces inflammation in vivo Owing to its characterized mechanism of modulating lipid mediator pathways it serves as a valuable tool for studying anti-inflammatory mechanisms and exploring new therapeutic strategies for inflammatory disorders
Catalog No. 50-000-95117