Learn More
CPC Scientific H-Met-Gln-Met-Lys-Lys-Val-Leu-Asp-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATIP001A
Encompass
SEQUENCE: H-Met-Gln-Met-Lys-Lys-Val-Leu-Asp-Ser-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: MQMKKVLDSMOLECULAR FORMULA: C45H82N12O14S2MOLECULAR WEIGHT: 1079.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [118850-71-8]SYNONYMS: RESEARCH AREA: InflammationREFERENCES:L.Miele et al., Nature, 335, 726 (1988)G.Camussi et al., J. Exp. Med. , 171, 913 (1990)V.Calderaro et al., J. Pharmacol. Exp. Ther. , 263, 579 (1992)A.B.Mukherjee, US Pat. Appl. 7-122 379 (Nov. 19) (1997)SKU(s): ATIP-001A
Catalog No. 50-210-8803