Learn More
Medchemexpress LLC Anti-Influenza A vir 1mg | 1mg
SDP
Catalog No. 5000324199
Encompass_Preferred
Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) is mouse-derived IgG2a type antibody inhibitor targeting to influenza virus nucleoprotein Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) reacts with influenza virus nucleoprotein (NP) Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) can induce passive immunity to influenza A virus Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) can be used for the detections of western blot flow cytometry and immunohistochemistry in Influenza A virus infection[1][2]
Catalog No. 50-003-24199 Supplier Medchemexpress LLC Supplier No. HYP9908111MG
Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) is mouse-derived IgG2a type antibody inhibitor targeting to influenza virus nucleoprotein Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) reacts with influenza virus nucleoprotein (NP) Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) can induce passive immunity to influenza A virus Anti-Influenza A virus NP Antibody (H16-L10-4R5 (HB-65)) can be used for the detections of western blot flow cytometry and immunohistochemistry in Influenza A virus infection[1][2]