Learn More
BIOHIPPO Anti-Influenza A virus Neuraminidase/NA Antibody (NC10), 100 µg
SDP
Catalog No. 5000859177
Encompass_Preferred
Influenza A virus Neuraminidase/NA Recombinant Antibody (clone NC10) is a recombinant Human IgG1 antibody specific for Influenza A virus Neuraminidase/NA. It reacts with Influenza A virus. Suitable for ELISA, SPR. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-59177 Supplier BIOHIPPO Supplier No. BHA21407625
Influenza A virus Neuraminidase/NA Recombinant Antibody (clone NC10) is a recombinant Human IgG1 antibody specific for Influenza A virus Neuraminidase/NA. It reacts with Influenza A virus. Suitable for ELISA, SPR. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.