Learn More
ECM Bioscience iNOS (C-terminal region) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience NP2131
Encompass_Preferred
The antibody detects a 130 kDa* protein on SDS-PAGE immunoblots of mouse macrophages (RAW264.7) treated with IFNγ and LPS, or J774A.1 cells treated with LPS only. The immunogen was Amino acid residues within the C-terminal region of human iNOS. The human iNOS sequence used has high homology with similar regions in rat and mouse iNOS. Accession #P35228. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Nos2 nitric oxide synthase 2, inducible, macrophage, NOS, type II, NOSII, Hepatocyte
Catalog No. 50-206-8768