Learn More
Sino Biological Anti-Integrin alpha 5 beta 1 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000785574
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Integrin alpha 5 beta 1 (rh Integrin alpha 5 beta 1; Catalog#CT014-H2508H; P08648 & P05556-1; Met1-Tyr995 & Met1-Asp728). Integrin alpha 5 beta 1 specific IgG was purified by Human Integrin alpha 5 beta 1 affinity chromatography.
Catalog No. 50-007-85574 Supplier Sino Biological Supplier No. CT014T16100
Produced in rabbits immunized with purified, recombinant Human Integrin alpha 5 beta 1 (rh Integrin alpha 5 beta 1; Catalog#CT014-H2508H; P08648 & P05556-1; Met1-Tyr995 & Met1-Asp728). Integrin alpha 5 beta 1 specific IgG was purified by Human Integrin alpha 5 beta 1 affinity chromatography.