Learn More
ECM Bioscience Integrin β3 (Extracellular region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience IM5811
Encompass_Preferred
Clone M581 was generated from a recombinant protein containing amino acid residues in the extracellular region of human Integrin β3. This sequence has high homology with rat and mouse Integrin β3, and has low homology to other integrin family members. Accession #P05106. This antibody detects a 100kDa* protein corresponding to the molecular mass of Integrin β3 on SDS-PAGE immunoblots of human platelets and endothelial cells (HUVEC). Applications: WB, ICC, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Integrin beta-3, Platelet membrane glycoprotein IIIa, GPIIIa, CD61, ITGB3, GP3A
Catalog No. 50-206-8733