Learn More
Sino Biological Anti-IRAK4 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000749988
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IRAK4 (rh IRAK4; Catalog#10735-H07B; Q9NWZ3-1; Met1-Ser460). IRAK4 specific IgG was purified by Human IRAK4 affinity chromatography.
Catalog No. 50-007-49988 Supplier Sino Biological Supplier No. 10735T32100
Produced in rabbits immunized with purified, recombinant Human IRAK4 (rh IRAK4; Catalog#10735-H07B; Q9NWZ3-1; Met1-Ser460). IRAK4 specific IgG was purified by Human IRAK4 affinity chromatography.