Learn More
Sino Biological Anti-IRAK4 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750969
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human IRAK4, and purified by antigen affinity chromatography.
Catalog No. 50-007-50969 Supplier Sino Biological Supplier No. 112647T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human IRAK4, and purified by antigen affinity chromatography.