Learn More
Sino Biological Anti-IRAK4 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750970
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human IRAK4, and purified by antigen affinity chromatography.
Catalog No. 50-007-50970 Supplier Sino Biological Supplier No. 112647T40100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human IRAK4, and purified by antigen affinity chromatography.