Learn More
ECM Bioscience JNK (Thr-183/Tyr-185), phospho-specific Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience JM2681
Encompass_Preferred
Clone M268 was generated from a dual phosphorylated peptide corresponding to amino acid residues surrounding Thr-183 and Tyr-185 in human JNK1. This sequence has high homology to the conserved site in rat and mouse JNK1 and JNK2. Accession #P45983. This antibody detects 46 and 54 kDa* proteins corresponding to the apparent molecular mass of JNK1 and JNK2 on SDS-PAGE immunoblots of human A431 and HeLa cells, as well as rat PC12 cells treated with calyculin A. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = SAPK1, p49MAPK, JNK1, JNK2, MAPK
Catalog No. 50-206-8743