Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-Kappa Light Chain (APC) TB28-2 ASR
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4048045
Encompass_Preferred
The CapricoBio Anti-Kappa Light Chain (APC) TB28-2 ASR, a monoclonal antibody for flow cytometry, targets human B cell kappa light chains. Conjugated to APC, it's used for immunophenotyping and comes in 100 tests of 50 μL each. Produced from TB28-2 hybridoma, it's affinity-purified and should be used at 5 μL/test, stored at 2-8°C, and shielded from light.
Catalog No. 50-227-9293