Learn More
Selleck Chemical LLC Anti-Klotho Beta-A2416-1MG
SDP
Catalog No. 503135370
Encompass_Preferred
Anti-Klotho Beta is an antibody targeting Beta Klotho (KLB) MW 145 24 kD
Catalog No. 50-313-5370 Supplier Selleck Chemical LLC Supplier No. A24161MG
Anti-Klotho Beta is an antibody targeting Beta Klotho (KLB) MW 145 24 kD