Learn More
PTM BIO LLC Anti-L-Lactyllysine Rabbit MAB, Catalog Number: PTM-1401RM, Rabbit, Recombinant Monoclonal, 12 months storage stability, Liquid
Catalog No. NC2099071
Encompass
The Anti-L-Lactyllysine Rabbit Monoclonal Antibody (PTM-1401RM) from PTM BIO LLC is a highly specific recombinant antibody aimed at detecting L-lactylated lysine residues in proteins. This pan-specific antibody uniquely recognizes its target independent of surrounding amino acid sequences, demonstrating no cross-reactivity with D-lactylated or N-ε-carboxyethylated peptides. It is an essential tool for researchers investigating the role of L-lactylation, a post-translational modification influenced by lactate metabolism with implications in gene expression and cancer biology.
  • Designed for the detection of L-lactylated lysine residues
  • Exhibits no cross-reactivity with unrelated modifications
  • Provides critical insights into post-translational modifications
  • Compatible with multiple applications such as WB, IHC, and FC
  • Stable for 12 months upon receipt and reconstitution
This item is not returnable. View return policy
Catalog No. NC2099071 Supplier PTM BIO LLC Supplier No. PTM1401RM
The Anti-L-Lactyllysine Rabbit Monoclonal Antibody (PTM-1401RM) from PTM BIO LLC is a highly specific recombinant antibody aimed at detecting L-lactylated lysine residues in proteins. This pan-specific antibody uniquely recognizes its target independent of surrounding amino acid sequences, demonstrating no cross-reactivity with D-lactylated or N-ε-carboxyethylated peptides. It is an essential tool for researchers investigating the role of L-lactylation, a post-translational modification influenced by lactate metabolism with implications in gene expression and cancer biology.
  • Designed for the detection of L-lactylated lysine residues
  • Exhibits no cross-reactivity with unrelated modifications
  • Provides critical insights into post-translational modifications
  • Compatible with multiple applications such as WB, IHC, and FC
  • Stable for 12 months upon receipt and reconstitution