Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyclonal Anti-Laminin Antibody | Not applicable | 0.5 mg/mL | Rabbit Source
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L9393.2ML
Encompass
"Anti-Laminin antibody | Purity: Not applicable | Mol Wt: 150 kDa | CAS No: Not applicable | Not applicable | 0.5 mg/mL" This polyclonal antibody was raised in rabbit against laminin, a major component of basement membranes, and is supplied as a buffered solution containing sodium azide preservative. The immunogen was laminin isolated from mouse sarcoma. This laminin antibody can be used for immunohistochemistry, immunocytochemistry, and ELISA assays. It shows reactivity with laminin from human, other mammals, birds, reptiles and amphibians. Keywords: laminin antibody, laminin IHC, laminin ICC, laminin ELISA, anti-laminin antibody, polyclonal laminin antibody, laminin immunocytochemistry.
Catalog No. NC2582540