Learn More
FUJIFILM Biosciences Anti Laminin Guinea Pig 50uL
Catalog No. 5000450111
Encompass_Preferred
Laminin is a component of the basement membrane an extracellular matrix and is a large protein with a heterotrimeric structure consisting of one alpha-chain one beta-chain and one gamma-chain Laminin is involved in many biological activities such as cell adhesion migration proliferation and promotion of axonal growth and differentiation through binding to integrins which are receptors for laminin This product is a guinea pig polyclonal antibody that recognizes Laminin Its available for a vessel marker Specificity to laminin
Catalog No. 50-004-50111 Supplier FUJIFILM Biosciences Supplier No. 01028821
Laminin is a component of the basement membrane an extracellular matrix and is a large protein with a heterotrimeric structure consisting of one alpha-chain one beta-chain and one gamma-chain Laminin is involved in many biological activities such as cell adhesion migration proliferation and promotion of axonal growth and differentiation through binding to integrins which are receptors for laminin This product is a guinea pig polyclonal antibody that recognizes Laminin Its available for a vessel marker Specificity to laminin