Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-LC3 antibody produced in rabbit, 200 uL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L8918200UL
Encompass
The gene LC3, also referred to as MAP1LC3A (microtubule associated protein 1 light chain 3 alpha), is a homologue of Apg8p necessary for autophagy in yeast. The cytosolic form upon processing gets localized to the autophagosome membranes. The gene is mapped to human chromosome 20q11.22. LC3 exists in cells as cytoplasmic LC3-I (18kDa) and LC3-II (16kDa).
Catalog No. NC1900441