Learn More
MyBioSource Anti-LILRB1 Antibody (PE), Mouse Monoclonal; 100 Test
SDP
Supplier:  MyBioSource MBS8107689
Encompass_Preferred
Anti-LILRB1 Antibody (PE), Mouse Monoclonal; 100 Tests. Specificity: Human CD85/ILT2/ILR1; Immunogen: Recombinant Human CD85/ILT2/ILR1 protein.For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2744046