Learn More
ECM Bioscience LIMK1 (C-terminus) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience LP1831
Encompass_Preferred
This antibody was affinity purified using LIMK1 (C-terminus) peptide (without carrier). The antibody detects a 72 kDa* protein corresponding to the molecular mass of LIMK1 on SDS-PAGE immunoblots of mouse recombinant LIMK1. The immunogen was LIMK1 (C-terminus) synthetic peptide (coupled to carrier protein) corresponding to amino acids at the C-terminus of human LIMK1. This sequence is conserved in rat and mouse LIMK1, and is not found in LIMK2. Accession #P53667. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = LIMK
Catalog No. 50-206-8750