Learn More
Alpco Ltd Anti-Liver-Kidney Microsomes-1 IgG ELISA (Anti-LKM-1 IgG ELISA)
SDP
Supplier:  Alpco Ltd 35LKMHUE01
Encompass_Preferred
The Anti-LKM-1 (Liver-Kidney Microsomes) (IgG) ELISA is a solid phase enzyme immunoassay employing human recombinant cytochrome p450 IID6 for the quantitative and qualitative detection of antibodies against liver-kidney microsomes (LKM) in human serum. The assay is a tool for the study of autoimmune hepatitis (AIH). Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2329140