Learn More
Sino Biological Anti-LOX-1 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000786300
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus OLR1 / LOX1 (rh OLR1 / LOX1; Catalog#90155-C07H; G7PJT2; Ser61-Gln273). OLR1 / LOX1 specific IgG was purified by Cynomolgus OLR1 / LOX1 affinity chromatography.
Catalog No. 50-007-86300 Supplier Sino Biological Supplier No. 90155T16100
Produced in rabbits immunized with purified, recombinant Cynomolgus OLR1 / LOX1 (rh OLR1 / LOX1; Catalog#90155-C07H; G7PJT2; Ser61-Gln273). OLR1 / LOX1 specific IgG was purified by Cynomolgus OLR1 / LOX1 affinity chromatography.