Learn More
Bio X Cell Antibody, Mouse, Rat IgG1, Unconjugated, 100 MG
Supplier: Bio X Cell BE0124R100MG
The MJ18 monoclonal antibody targets mouse CD276 (B7-H3), a type I transmembrane protein involved in T cell responses. This antibody is designed for in vivo B7-H3 blockade and flow cytometry applications.
- Clone: MJ18
- Host species: Rat
- Isotype: Rat IgG1, κ
- Reacts with mouse CD276 (B7-H3)
- Applications include in vivo B7-H3 blockade and flow cytometry
- Purity: ≥95% by SDS-PAGE
- Unconjugated
- Immunogen: Mouse B7-H3 IgG2a fusion protein
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtered
- Purified from cell culture supernatant in an animal-free facility
- Purification method: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.