Learn More
Bio X Cell Monoclonal Antibody, BTLA, Clone PJ196, 5 MG
Supplier: Bio X Cell BE0196R005MG
The PJ196 monoclonal antibody reacts with mouse B- and T-lymphocyte attenuator (BTLA), also known as CD272. BTLA is an Ig superfamily member expressed on various immune cells and interacts with B7 homolog B7-H4 and herpes virus entry mediator (HVEM), negatively regulating T cell immune responses.
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Sterility: 0.2 µm filtered
- Purified from cell culture supernatant in an animal-free facility
- Storage: Store at 4°C; do not freeze
- Molecular weight: 150 kDa
- Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
- Reported applications: In vivo BTLA blockade, in vitro T cell stimulation/activation, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.