Learn More
Bio X Cell Antibody, CSF1R (CD115) AFS98 IgG2a, 100 MG
Supplier: Bio X Cell BE0213R100MG
The AFS98 monoclonal antibody reacts with mouse colony stimulating factor 1 receptor (CSF1R), also known as macrophage colony-stimulating factor receptor (M-CSFR), and CD115. CSF1R is a single-pass type I membrane protein and a member of the platelet-derived growth factor receptor family. This antibody has been reported to deplete macrophages and block CSFR1 in vivo.
- Isotype: Rat IgG2a, κ
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Purification: Protein G
- Production: Purified from cell culture supernatant in an animal-free facility
- Reported applications: In vivo macrophage depletion, in vitro CSF1R neutralization, in vivo monocyte depletion, Flow cytometry, Western blot
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.