Learn More
Bio X Cell anti-mouse CSF1R (CD115) Antibody, AFS98, 25 MG
Supplier: Bio X Cell BE0213R025MG
The AFS98 monoclonal antibody targets mouse colony stimulating factor 1 receptor (CSF1R), also known as macrophage colony-stimulating factor receptor (M-CSFR), and CD115. This antibody is a single-pass type I membrane protein receptor for CSF1, regulating proliferation and differentiation of cells in the monocytic lineage. It depletes macrophages and blocks CSFR1 in vivo.
- Isotype: Rat IgG2a, κ
- Purity: ≥95% (determined by SDS-PAGE)
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin: ≤1EU/mg (determined by LAL assay)
- Sterility: 0.2 μm filtration
- Production: Purified from animal-free cell culture supernatant
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
- Applications: In vivo macrophage depletion, in vitro CSF1R neutralization, flow cytometry, western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.