Learn More
Bio X Cell Antibody, Anti-mouse OX40, Clone OX-86, 25 MG
Supplier: Bio X Cell BE0031R025MG
The OX-86 monoclonal antibody targets mouse OX-40 (CD134), a 50 kDa type I membrane glycoprotein and a member of the TNF receptor superfamily. It is expressed on activated CD4 and CD8 T cells, regulatory T cells, NKT cells, NK cells, and neutrophils. This agonistic antibody has been shown to delay tumor growth in vivo.
- Clone: OX-86
- Target: Mouse OX40 (CD134)
- Host species: Rat
- Isotype: Rat IgG1, κ
- Reactivities: Mouse
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Storage: Store at stock concentration at 4°C; do not freeze
- Applications: In vivo OX40 activation, in vitro OX40 activation, Western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.