Learn More
Bio X Cell Antibody, Anti-Mouse CTLA-4, Clone 9H10, 25 MG
Supplier: Bio X Cell BE0131R025MG
The 9H10 monoclonal antibody reacts with mouse CTLA-4 (CD152), a 33 kDa cell surface receptor expressed on activated T and B lymphocytes. It negatively regulates immune responses and is a key target in cancer immunotherapy via immune checkpoint blockade.
- Isotype: Syrian hamster IgG
- Reactivities: Mouse
- Conjugation: Unconjugated
- Immunogen: Mouse CTLA-4-human IgG1 fusion protein
- Reported applications: in vivo CTLA-4 neutralization, in vitro CTLA-4 neutralization, Western blot
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.