Learn More
Bio X Cell Antibody, Mouse CTLA-4, Hamster IgG, 5 MG
Supplier: Bio X Cell BE0131R005MG
InVivoMAb anti-mouse CTLA-4 (CD152) is an antibody solution provided in a 5 mg package, designed for various in vivo and in vitro applications.
- Applications: in vivo CTLA-4 neutralization, in vitro CTLA-4 neutralization, Western blot
- Antibody host: Syrian Hamster
- Species reactivity: Mouse
- Antibody isotype: IgG
- Immunogen: Mouse CTLA-4-human IgG1 fusion protein
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.