Learn More
Bio X Cell InVivoMAb anti-mouse CTLA-4 (UC10-4F10-11), 100 MG
Supplier: Bio X Cell BE0032R100MG
The UC10-4F10-11 monoclonal antibody reacts with mouse CTLA-4 (cytotoxic T lymphocyte antigen-4), also known as CD152. This antibody negatively regulates cell-mediated immune responses and promotes T cell co-stimulation by blocking CTLA-4 binding to B7 co-receptors.
- Formulation: PBS, pH 6.5, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Validated applications: In vivo CTLA-4 neutralization, in vitro CTLA-4 neutralization, flow cytometry, western blot
- Unconjugated format
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.