Learn More
Bio X Cell Monoclonal Antibody, Mouse CD19, 1D3, 5 MG
Supplier: Bio X Cell BE0150R005MG
The 1D3 monoclonal antibody reacts with mouse CD19, a B cell-specific 95 kDa transmembrane glycoprotein. It functions as a positive regulator of B-cell receptor signaling and is highly expressed in lymphomas and leukemias. This antibody is reported to deplete B cells and is an attractive target for immunotherapy applications.
- Reacts with mouse CD19
- Monoclonal antibody, clone 1D3
- Unconjugated
- Endotoxin ≤1 EU/mg
- Purity ≥95%
- Sterility: 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility
- Applications include in vivo B cell depletion, in vivo CD19 neutralization, in vitro B cell negative selection, and flow cytometry
- Purification method: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.