Learn More
Bio X Cell anti-mouse CD19 Antibody, 1D3, IgG2a, 25 MG
Supplier: Bio X Cell BE0150R025MG
The 1D3 monoclonal antibody reacts with mouse CD19, a B cell-specific 95 kDa transmembrane glycoprotein. CD19 regulates B-cell receptor signaling and is highly expressed in many lymphomas and leukemias, making it an attractive target for immunotherapy.
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Purified in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.