Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD1d, Clone 20H2, Rat IgG1, 5 MG
Supplier: Bio X Cell BE0179R005MG
The 20H2 monoclonal antibody reacts with mouse CD1d, also known as CD1.1. CD1d is a 48 kDa type I membrane glycoprotein and a member of the CD1 family of glycoproteins. It is a non-classical MHC protein with structural homology to class I MHC molecules. CD1d is expressed on the surface of various antigen-presenting cells and is involved in the presentation of non-peptide glycolipid antigens to CD1d-restricted T cells. CD1d-presented glycolipid antigens activate invariant natural killer T (iNKT) cells, which then rapidly produce Th1 and Th2 cytokines.
- Reacts with mouse CD1d (CD1.1)
- Clone 20H2 (HB323)
- Isotype: Rat IgG1,
- Unconjugated
- Formulated in PBS, pH 7.0
- Contains no stabilizers or preservatives
- Endotoxin level of 1EU/mg
- Purity of 95%
- Sterilized by 0.2 m filtration
- Produced in an animal-free facility
- Purified by protein G
- Supports iNKT cell neutralization
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.