Learn More
Bio X Cell Antibody, Mouse CD209b, Clone 22D1, 5 MG
Supplier:  Bio X Cell BE0220R005MG
Encompass_Preferred
The 22D1 monoclonal antibody reacts with mouse CD209b, also known as SIGN-R1. CD209b is a 37 kDa type II transmembrane C-type lectin receptor. It is expressed on the surface of splenic marginal zone and lymph node medullary macrophages and serves as a marker for these cells. CD209b is involved in the innate immune response, binding to and initiating the uptake of various microorganisms by recognizing high-mannose-containing glycoproteins on their envelopes. The 22D1 antibody has been reported to block CD209b in vivo.
  • Monoclonal antibody 22D1 reacts with mouse CD209b (SIGN-R1).
  • CD209b is a 37 kDa type II transmembrane C-type lectin receptor.
  • Expressed on splenic marginal zone and lymph node medullary macrophages.
  • Involved in innate immune response.
  • Binds to and initiates uptake of microorganisms via high-mannose-containing glycoproteins.
  • Antibody 22D1 blocks CD209b in vivo.
Catalog No. 50-139-3241