Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD25, PC-61.5.3, 0.25 MG
Supplier: Bio X Cell BE0012R025MG
This monoclonal antibody, clone PC-61.5.3, reacts with mouse IL-2Rα (CD25), the 55 kDa ligand-binding subunit of the interleukin 2 receptor alpha chain. It is commonly used to deplete CD4+FoxP3+ T regulatory cells in vivo and inhibits IL-2 binding to both low and high affinity IL-2 receptor forms.
- Clone: PC-61.5.3
- Isotype: Rat IgG1, λ
- Host species: Rat
- Applications: In vivo regulatory T cell depletion, flow cytometry
- Purity: >95% by SDS-PAGE
- Sterility: 0.2 μM filtered
- Endotoxin: <2EU/mg (<0.002EU/μg)
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.