Learn More
Bio X Cell Mouse CD28 Antibody 37.51, Unconjugated, 25 MG
Supplier: Bio X Cell BE00151R025MG
The 37.51 monoclonal antibody targets mouse CD28, a 45 kDa costimulatory receptor found on thymocytes, most peripheral T cells, and NK cells. It acts as a receptor for CD80 (B7-1) and CD86 (B7-2) and enhances IL-2 and IL-2 receptor expression, increases the cytotoxicity of CD3-activated T cells, and provides a costimulatory signal for CTL induction.
- Isotype: Syrian Hamster IgG2
- Immunogen: C57BL/6 mouse T cell lymphoma EL-4 cells
- Applications: in vitro T cell stimulation/activation, in vivo CD28 blockade
- Formulation: PBS, pH 6.0; 0.01% Tween; no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtered
- Production: Purified from animal-free cell culture supernatant
- Purification: Protein G
- Molecular Weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.