Learn More
Bio X Cell Anti-Mouse CD28, Clone 37.51, IgG2, 5 MG
Supplier: Bio X Cell BE00151R005MG
The 37.51 monoclonal antibody reacts with mouse CD28, a 45 kDa costimulatory receptor and a member of the Ig superfamily. CD28 is expressed by thymocytes, most peripheral T cells, and NK cells. It augments IL-2 and IL-2 receptor expression, and stimulates proliferation and cytokine production by activated T and NK cells, providing a costimulatory signal for CTL induction.
- Clone: 37.51
- Reactivity: Mouse
- Isotype: Syrian Hamster IgG2
- Immunogen: C57BL/6 mouse T cell lymphoma EL-4 cells
- Formulation: PBS, pH 6.0, 0.01% Tween, no stabilizers/preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtered
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.