Learn More
Bio X Cell Antibody, Mouse CD3e, Clone 145-2C11, 0.25 MG
Supplier:  Bio X Cell BE00011R025MG
Encompass_Preferred
The 145-2C11 monoclonal antibody targets mouse CD3ε, a 20 kDa transmembrane cell-surface protein belonging to the immunoglobulin superfamily. It plays roles in TCR signaling, T lymphocyte activation, and antigen recognition. This antibody induces T lymphocyte activation, proliferation, and apoptosis by binding and stimulating the TCR.
  • Reacts with mouse
  • Induces T lymphocyte activation, proliferation, and apoptosis
  • Blocks binding of the 17A2 antibody to CD3ε+ T lymphocytes
  • Unconjugated
  • Purified from cell culture supernatant in an animal-free facility
  • Has ≥95% purity as determined by SDS-PAGE
  • Has ≤1EU/mg endotoxin (≤0.001EU/μg)
Catalog No. 50-561-953