Learn More
Bio X Cell Antibody, Mouse CD3, Clone 145-2C11, 5 MG
Supplier: Bio X Cell BE00011R005MG
The 145-2C11 monoclonal antibody reacts with mouse CD3ε, a 20 kDa transmembrane cell-surface protein of the immunoglobulin superfamily. CD3ε forms part of the TCR complex and is expressed on T lymphocytes, NK-T cells, and developing thymocytes. This antibody induces T lymphocyte activation, proliferation, and apoptosis by binding and stimulating the TCR.
- Clone: 145-2C11
- Host: Armenian hamster
- Target: Mouse CD3ε
- Isotype: Armenian hamster IgG1
- Applications: In vivo T cell depletion, in vitro T cell activation, immunofluorescence, flow cytometry, western blot
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purity: ≥95%
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.