Learn More
Bio X Cell anti-mouse CD4 Antibody, Clone GK1.5, Rat IgG2b, Unconjugated, 25 MG
Supplier: Bio X Cell BP00031R025MG
The GK1.5 monoclonal antibody specifically targets mouse CD4, a 55 kDa cell surface glycoprotein vital for T cell development and optimal function. It acts as a co-receptor, interacting with class II MHC molecules on antigen-presenting cells. This antibody is expressed by most thymocytes and helper T cells, making it suitable for a range of immunological research applications.
- In vivo CD4+ T cell depletion
- Flow cytometry and western blot compatible
- Reacts with mouse CD4
- Isotype: Rat IgG2b,
- Purity: 95% by SDS-PAGE
- Endotoxin level: 0.5EU/mg
- Aggregation level: <5%
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Unconjugated (conjugation services available)
- Purified in an animal-free facility using Protein G
- Molecular weight: 150 kDa
- Tested negative for murine pathogens
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.