Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD4, GK1.5, 25 MG
Supplier: Bio X Cell BE00031R025MG
The GK1.5 monoclonal antibody reacts with the mouse CD4, a 55 kDa cell surface glycoprotein. CD4 acts as a co-receptor with the T cell receptor (TCR), interacting with class II MHC molecules on antigen presenting cells (APC). CD4 is expressed by thymocytes, helper T cells, NK-T cells, dendritic cells, and macrophages, playing a crucial role in T cell development and function.
- In vivo CD4+ T cell depletion
- Flow cytometry
- Western blot
- Purified from cell culture supernatant in an animal-free facility
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg determined by LAL assay
- Purity: ≥95% determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular Weight: 150 kDa
- Purification: Protein G
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.