Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD4, GK1.5, 50 MG
Supplier: Bio X Cell BE00031R050MG
The GK1.5 monoclonal antibody reacts with mouse CD4, a 55 kDa cell surface glycoprotein. CD4 acts as a co-receptor with the T cell receptor (TCR), interacting with class II MHC molecules on antigen presenting cells (APC).
- Isotype: Rat IgG2b, κ
- Conjugation: Unconjugated
- Immunogen: Mouse CTL clone V4
- Reported applications: In vivo CD4+ T cell depletion, flow cytometry, western blot
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.