Learn More
Bio X Cell Anti-Mouse CD4, GK1.5, Rat IgG2b, 100 MG
Supplier: Bio X Cell BE00031R100MG
The GK1.5 monoclonal antibody reacts with mouse CD4, a 55 kDa cell surface glycoprotein essential for T cell development and optimal function. It acts as a co-receptor with the T cell receptor, interacting with class II MHC molecules on antigen-presenting cells.
- Clone: GK1.5
- Host species: Rat
- Target species: Mouse
- Isotype: Rat IgG2b, κ
- Applications: In vivo CD4+ T cell depletion, flow cytometry, western blot
- Conjugation: Unconjugated
- Purity: ≥95%
- Endotoxin: ≤1EU/mg
- Formulation: PBS, pH 6.5
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.