Learn More
Bio X Cell Anti-Mouse CD4, Clone GK1.5, Rat, 5 MG
Supplier: Bio X Cell BE00031R005MG
This InVivoMAb anti-mouse CD4 antibody is designed for various immunology applications. It features high purity and ultra-low endotoxin levels, ensuring reliable performance in sensitive assays.
- Host species: rat
- Target species: mouse
- Clone: GK1.5
- Antibody isotype: IgG2b, κ
- Applications: in vivo CD4+ T cell depletion, flow cytometry, western blot
- Size: 5 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.