Learn More
Bio X Cell Antibody, anti-mouse CD4, YTS 177, 5 MG
Supplier: Bio X Cell BE00033R005MG
The YTS 177 monoclonal antibody reacts with the mouse CD4 antigen, a 55 kDa cell surface type I membrane glycoprotein crucial for T cell development and optimal function. It acts as a co-receptor with the T cell receptor to interact with class II MHC molecules on antigen-presenting cells. The antibody is non-depleting but induces rapid internalization of CD4.
- Reacts with mouse CD4 antigen, a 55 kDa glycoprotein
- Acts as co-receptor with T cell receptor (TCR)
- Interacts with class II MHC molecules on antigen-presenting cells
- Non-depleting; induces rapid internalization of CD4
- Suppresses allograft rejection, allergic reactions, and autoimmune responses
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1EU/mg
- Sterility: 0.2 μm filtration
- Purification: Protein G
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.