Learn More
Bio X Cell Anti-mouse CD40, FGK4.5, Rat IgG2a, 5 MG
Supplier:  Bio X Cell BP00162R005MG
Encompass_Preferred
The FGK4.5 monoclonal antibody reacts with mouse CD40, a 48 kDa type I transmembrane glycoprotein expressed on APCs and other cells. CD40 acts as a costimulatory molecule for the activation of various immune cells and is involved in B cell activation, differentiation, proliferation, and Ig isotype switching. This agonistic antibody activates CD40 expressing APCs and can inhibit CD40/CD154 interaction in vitro and in vivo.
  • Agonistic antibody
  • Activates CD40 expressing APCs
  • Can inhibit CD40/CD154 interaction in vitro and in vivo
Catalog No. 50-139-3517