Learn More
Bio X Cell anti-mouse CD40 FGK4.5/FGK45, Rat IgG2a, 25 MG
Supplier: Bio X Cell BE00162R025MG
The FGK4.5 monoclonal antibody targets mouse CD40, a 48 kDa type I transmembrane glycoprotein of the TNF receptor superfamily. This agonistic antibody activates CD40 expressing antigen-presenting cells (APCs) and can also inhibit CD40/CD154 interaction.
- Targets mouse CD40 (Bp50)
- Clone FGK4.5/FGK45, host rat, isotype rat IgG2a
- For in vivo CD40 activation and in vitro B cell stimulation/activation
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin ≤1 EU/mg, purity ≥95%
- Purified via Protein G from animal-free facility
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.